
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester 95%
Catalog # H33958.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:170842-80-5
:
Formula:C₂₀H₂₉NO₄
MDL Number:MFCD11500859
:347.46 g/mol
:GHS07
Storage Temperature:Ambient