3-{4-(Methylsulphonyl)benzene}-1-propanoic acid 96%
Catalog # H33764.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.27 g/mol
Melting Pt:173…177 °C
:GHS07
Storage Temperature:Ambient
CAS Number:387350-46-1
Formula:C₁₀H₁₂O₄S
MDL Number:MFCD01319236