
3'-Methoxybiphenyl-4-sulphonyl chloride 96%
Catalog # H33211.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
CAS Number:186550-26-5
:
Formula:C₁₃H₁₁ClO₃S
MDL Number:MFCD06409270
:282.74 g/mol
Melting Pt:80…84 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05