Order Entry
Northern Ireland
ContactUsLinkComponent
3-(3,5-Di-tert-butylphenyl)propionic acid 96%
Irritant
3-(3,5-Di-tert-butylphenyl)propionic acid 96%
Catalog # H33183.ME
CAS Number:  
Irritant
3-(3,5-Di-tert-butylphenyl)propionic acid 96%
Catalog # H33183.ME
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

CAS Number:42288-01-7
 :
Formula:C₁₇H₂₆O₂
MDL Number:MFCD09258906
 :262.39 g/mol
Melting Pt:103…108 °C
 :GHS07
Storage Temperature:Ambient