
Ethyl-2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate 96%
Catalog # H33171.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:281.76 g/mol
Melting Pt:71…73 °C
:GHS07
Storage Temperature:Ambient
CAS Number:325724-66-1
Formula:C₁₃H₁₂ClNO₂S
MDL Number:MFCD01924157