Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole 95%
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole 95%
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole 95%
H31755.MD
:
Thermo Fisher Scientific
:
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole 95%
H31755.MD
:
Thermo Fisher Scientific
:
Pk:
250 mg
Seq:
2590
:
478828-53-4
:
:
C₁₂H₁₃BrN₂O
:
MFCD12922964
:
281.15 g/mol
:
GHS07
:
Ambient