N-Boc-4-oxo-L-proline methyl ester 97%
Catalog # H27782.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:243.259 g/mol
Melting Pt:35…40 °C
CAS Number:102195-80-2
Formula:C₁₁H₁₇NO₅
MDL Number:MFCD01861778