Order Entry
Northern Ireland
ContactUsLinkComponent
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
Catalog # H27774.03
CAS Number:  
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
Catalog # H27774.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:1194-16-7
 :
Formula:C₇H₁₂O₂
 :128.17 g/mol
Boiling Pt:60…64 °C (8 mmHg)
MDL Number:MFCD01549337