
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II)
Catalog # H26891.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:12289-94-0
:
Formula:C₁₆H₂₆Ru
MDL Number:MFCD00216965
:319.454 g/mol
Melting Pt:83…86 °C
ADR:4.3,III
:GHS02