Order Entry
Northern Ireland
ContactUsLinkComponent
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
Catalog # H26882.03
CAS Number:  
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene ≥99%
Catalog # H26882.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:22692-80-4
 :
Formula:C₁₇H₁₆
 :220.315 g/mol
Boiling Pt:135…136 °C (1 mmHg)
Melting Pt:71…74 °C
Storage Temperature:Ambient
MDL Number:MFCD04038793