
2-Methoxy-4-(trifluoromethoxy)benzaldehyde 97%
Catalog # H26520.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Formula:C₉H₇F₃O₃
MDL Number:MFCD04115965
:220.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:886500-13-6