Order Entry
Northern Ireland
ContactUsLinkComponent
5-Fluoro-2-nitrophenylacetonitrile 99%
Toxic
Irritant
5-Fluoro-2-nitrophenylacetonitrile 99%
Catalog # H26125.03
CAS Number:  
Toxic
Irritant
5-Fluoro-2-nitrophenylacetonitrile 99%
Catalog # H26125.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:3456-75-5
 :
Formula:C₈H₅FN₂O₂
MDL Number:MFCD00039742
 :180.14 g/mol
Melting Pt:68…72 °C
ADR:6.1,III
 :GHS06
 :GHS07
Storage Temperature:Ambient