
2-Phenylbenzimidazole-5-sulphonic acid monohydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27503-81-7
:
Formula:C₁₃H₁₀N₂O₃S·H₂O
MDL Number:MFCD00150559
:292.32 g/mol (274.30 anhy)
Melting Pt:>300 °C
EINECS:248-502-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together