


Zirconium(IV) butoxide ≥80% (w/w) in 1-butanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1071-76-7
:
Formula:C₁₆H₃₆O₄Zr
MDL Number:MFCD00048765
:383.68 g/mol
Density:1,070
Flash Pt:38 °C (100 °F)
EINECS:213-995-3
UN:1120
ADR:3,III
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together