Order Entry
Northern Ireland
ContactUsLinkComponent
3-Methyl-2-butanethiol 95% remainder mainly 2-methyl-1-butanethiol
Flammable
Irritant
3-Methyl-2-butanethiol 95% remainder mainly 2-methyl-1-butanethiol
Catalog # B22330.14
CAS Number:  
Flammable
Irritant
3-Methyl-2-butanethiol 95% remainder mainly 2-methyl-1-butanethiol
Catalog # B22330.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :104.21 g/mol
Boiling Pt:100 °C
Density:0,840
Flash Pt:3 °C (37 °F)
EINECS:218-223-9
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:2084-18-6
Formula:C₅H₁₂S
MDL Number:MFCD00039648