Malonaldehydebis(phenylimine) monohydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123071-42-1
:
Formula:C₁₅H₁₄N₂·HCl
MDL Number:MFCD00054634
Melting Pt:∼ 215 °C (dec.)
:258.75 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together