6-Chloro-3-indolyl-β-D-galactopyranoside 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:138182-21-5
:
Formula:C₁₄H₁₆ClNO₆
MDL Number:MFCD00467206
:329.733 g/mol
Melting Pt:188…190 °C
Related Information
Frequently Bought Together