Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Diphenyl-2-oxazolethiol
4,5-Diphenyl-2-oxazolethiol 98%
4,5-Diphenyl-2-oxazolethiol 98%
B20111.14
:
Thermo Fisher Scientific
:
4,5-Diphenyl-2-oxazolethiol 98%
B20111.14
:
Thermo Fisher Scientific
:
Pk:
25 g
Seq:
3308
:
6670-13-9
:
:
C₁₅H₁₁NOS
:
MFCD00173660
:
251…256 °C
:
253.32 g/mol
:
229-707-4
:
GHS07
:
Ambient
:
MFCD00022414