Order Entry
Northern Ireland
ContactUsLinkComponent
2,8-Bis(trifluoromethyl)quinolin-4-ol 97%
Irritant
2,8-Bis(trifluoromethyl)quinolin-4-ol 97%
Catalog # A19750.06
CAS Number:  
Irritant
2,8-Bis(trifluoromethyl)quinolin-4-ol 97%
Catalog # A19750.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:35853-41-9
 :
Formula:C₁₁H₅F₆NO
MDL Number:MFCD00075091
Melting Pt:129…135 °C
 :281.16 g/mol
EINECS:252-762-0
 :GHS07
Storage Temperature:Ambient