3-Nonen-2-one 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14309-57-0
:
Formula:C₉H₁₆O
MDL Number:MFCD00010241
Boiling Pt:85 °C (12 mmHg)
Density:0,848
Flash Pt:81 °C (177 °F)
:140.23 g/mol
Storage Temperature:Ambient
EINECS:238-248-9
Related Information
Frequently Bought Together