Methyl-3-(dimethylamino)-2-methylpropanoate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10205-34-2
:
Formula:C₇H₁₅NO₂
MDL Number:MFCD00044842
Boiling Pt:152…154 °C
Molecular Weight:145.2 g/mol
EINECS:233-505-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together