Order Entry
Northern Ireland
ContactUsLinkComponent
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 99%
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 99%
Catalog # A18696.06
CAS Number:  
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde 99%
Catalog # A18696.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :164.16 g/mol
Boiling Pt:104…106 °C (15 mmHg)
Melting Pt:48…52 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:249-763-3
CAS Number:29668-44-8
Formula:C₉H₈O₃
MDL Number:MFCD00010092