
8-Ethoxyquinoline-5-sulphonic acid hydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15301-40-3
:
Formula:C₁₁H₁₁NO₄S
MDL Number:MFCD00065059
:253.27 g/mol
Melting Pt:∼ 296 °C (dec.)
EINECS:239-334-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together