Phenyl trans-styryl sulphone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16212-06-9
:
Formula:C₁₄H₁₂O₂S
MDL Number:MFCD00159177
Melting Pt:74…76 °C
:244.31 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together