
Chloromethylphosphonothioic dichloride 98%
Catalog # A18456.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:183.42 g/mol
Boiling Pt:88…90 °C (30 mmHg)
ADR:8,II
:GHS05
Storage Temperature:Ambient
CAS Number:1983-27-3
Formula:CH₂Cl₃PS
MDL Number:MFCD00015876