Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Tyrosine ethyl ester hydrochloride
L-Tyrosine ethyl ester hydrochloride 99%
L-Tyrosine ethyl ester hydrochloride 99%
A18358.14
:
Thermo Fisher Scientific
:
L-Tyrosine ethyl ester hydrochloride 99%
A18358.14
:
Thermo Fisher Scientific
:
Pk:
25 g
Seq:
3308
:
4089-07-0
:
:
C₁₁H₁₅NO₃
:
MFCD00063047
:
245.70 g/mol
:
166…168 °C
:
223-820-2
:
GHS07
:
Ambient