
trans,cis-2,6-Nonadienal ≥90% (major isomer) remainder mainly trans,trans-isomer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:557-48-2
:
Formula:C₉H₁₄O
MDL Number:MFCD00007009
:138.21 g/mol
Boiling Pt:90 °C (4 mmHg)
Density:0,866
Flash Pt:76 °C (169 °F)
EINECS:209-178-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together