Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
2,5-Dithiobiurea 97%

2,5-Dithiobiurea 97%

Irritant
2,5-Dithiobiurea 97%
Certificates
2,5-Dithiobiurea 97%
2,5-Dithiobiurea 97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 142-46-1 Molecular Formula: C2H6N4S2 Formula Weight: 150.22 Melting Point: ca 212° dec. Beilstein Reference: 1723178 MDL No.: MFCD00014471

Synonyms:  Hydrazine-1,2-bis(carbothioamide)
  • CAS Number:142-46-1
  • Formula:C₂H₆N₄S₂
  • MDL Number:MFCD00014471
  • Molecular Weight:150.22 g/mol
  • Melting Pt:∼ 212 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:205-537-6
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £28.70
    £28.70
    1 * 25 g
       £28.70

  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £90.70
    £90.70
    1 * 100 g
       £90.70