Chromium(III)-2-ethylhexanoate 50% in 2-ethylhexanoic acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3444-17-5
:
Formula:C₂₄H₄₅CrO₆
MDL Number:MFCD00014005
:481.607 g/mol
EINECS:222-357-3
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together