Polyethylene oxide, M.W. >5,000,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25322-68-3
:
Formula:(-CH₂CH₂O-)n
MDL Number:MFCD00147970
Melting Pt:∼ 175 °C
Storage Temperature:Ambient
EINECS:500-038-2
Related Information
Frequently Bought Together