6-Acetyl-1,2,3,4-tetrahydronaphthalene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:774-55-0
:
Formula:C₁₂H₁₄O
MDL Number:MFCD00019710
:174.24 g/mol
Boiling Pt:119…121 °C (2 mmHg)
Melting Pt:–21 °C
Density:1,058
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:212-266-7
Related Information
Frequently Bought Together