
Azure II
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37247-10-2
:
Formula:C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
MDL Number:MFCD00080716
:625.68 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together