Iron(III) hydroxide, α-phase 20% in water stabilised, nanoparticle dispersion, Ø <5.0 nm APS Number Weighted
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20344-49-4
:
Formula:FeO(OH)
MDL Number:MFCD00064782
:88.85 g/mol
Merck Index:14,4025
Density:1,20
:Moistened with water
EINECS:243-746-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together