
Potassium bis(trifluoromethylsulphonyl)imide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90076-67-8
:
Formula:C₂F₆KNO₄S₂
MDL Number:MFCD06200829
:319.234 g/mol
Melting Pt:198…203 °C
Storage Temperature:Ambient
ADR:8,II
:GHS05
Related Information
Frequently Bought Together