
Bisphenol F acetate propionate epoxy curative EC-298
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:C₁₈H₁₈O₄
MDL Number:MFCD22380705
:298.34 g/mol
Density:1,13
Flash Pt:150 °C (302 °F)
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together