
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride (16,3% Pd)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95408-45-0
:
Formula:C₂₆H₄₄Cl₂FeP₂Pd
MDL Number:MFCD08064219
:651.75 g/mol
:Packaged under argon.
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together