Calcium-D gluconate 2,5% (w/w) in aqueous solution, gel
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:299-28-5
:
Formula:HOH₂C(CH(OH))₄COOCa
MDL Number:MFCD00064209
:430.38 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together