Poly(styrene) atactic, M.W. 125,000-250,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-53-6
:
Formula:C₈H₈
MDL Number:MFCD00084450
:125000 - 250000 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together