Poly(styrene-divinylbenzene), aminomethylated, cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89551-24-6
:
MDL Number:MFCD00146442
Storage Temperature:Ambient
Related Information
Frequently Bought Together