Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9016-00-6
:
Formula:(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
MDL Number:MFCD00148360
Boiling Pt:85…105 °C (2…4 mmHg)
Melting Pt:–40 °C
Density:0,88
Flash Pt:70 °C (158 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together