Order Entry
Northern Ireland
ContactUsLinkComponent
Rhodium 1% on polyethylenimine/ SiO₂, powder
Health
Rhodium 1% on polyethylenimine/ SiO₂, powder
Catalog # 43409.14
CAS Number:  
Health
Rhodium 1% on polyethylenimine/ SiO₂, powder
Catalog # 43409.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :102.9 g/mol
EINECS:231-125-0
 :GHS08
Storage Temperature:Ambient
CAS Number:7440-16-6
Formula:Rh
MDL Number:MFCD00011201