Sodium nitrite standardised solution 0.1 M in aqueous solution volumetric solution
500 mL
Supplier: Thermo Fisher Scientific
Sodium nitrite standardised solution 0.1 M in aqueous solution volumetric solution
Catalog # 43015.AP
Supplier: Thermo Fisher Scientific
CAS Number: 7632-00-0
Jump to:
Product Details & Documents
Formula Weight: 69.00 Physical Form: Liquid Appearance: Colourless Odour: Odourless
Synonyms: Diazotizing salts
- CAS Number:7632-00-0
- Formula:NaNO₂
- MDL Number:MFCD00011118
- Molecular Weight:69 g/mol
- Storage Temperature:Ambient
- EINECS:231-555-9
- UN:3287
- ADR:6.1,III
Documents
Specifications
Specifications
- Pk:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



