Order Entry
Northern Ireland
ContactUsLinkComponent
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 90.0 micron
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 90.0 micron
Catalog # 42751.AA
CAS Number:  
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 90.0 micron
Catalog # 42751.AA
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Ambient
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450