Order Entry
Northern Ireland
ContactUsLinkComponent
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.50 micron
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.50 micron
Catalog # 42714.4Y
CAS Number:  
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.50 micron
Catalog # 42714.4Y
CAS Number:  

Specifications

  • Pk:
    15 mL
  • Seq:
    2295

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Refrigerator
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450