Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9016-00-6
:
Formula:(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
MDL Number:MFCD00148360
Boiling Pt:>205 °C
Melting Pt:< –40 °C
Density:0,968
Flash Pt:315 °C (599 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together