
(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol ≥97%
Catalog # 42183.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.208 g/mol
Melting Pt:112…115 °C
EINECS:248-867-6
:GHS07
Storage Temperature:Ambient
CAS Number:28143-91-1
Formula:C₉H₁₃NO₂
MDL Number:MFCD00004503