Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Specimens
Mineral Specimens
Gems and Tumbled Stones
Ruby spheres
Ruby spheres
Ruby spheres
42076.KJ
:
Thermo Fisher Scientific
Ruby spheres
42076.KJ
:
Thermo Fisher Scientific
Ø:
1,25 mm
Grade:
25
Description:
Ruby spheres
Seq:
11
MDL No.:
MFCD00003424
Pk:
5
MDL No.: MFCD00003424