Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 1 mm (0.04 in) Type 316
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:Fe
MDL Number:MFCD00148499
:55.845 g/mol
Storage Temperature:Ambient
Frequently Bought Together