Polyvinylpyrrolidone cross-linked
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25249-54-1
:
Formula:C₆H₉NO
MDL Number:MFCD00149016
:111.144 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together