Tungsten zirconium oxide ≥99.7% (metals basis excluding Hf) (max. 0.3% Hf)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16853-74-0
:
Formula:ZrW₂O₈
MDL Number:MFCD00168096
:586.92 g/mol
Density:5,10
Storage Temperature:Ambient
EINECS:240-876-3
Related Information
Frequently Bought Together